Assay Details
| Gene Symbol: EGLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735291.3 | XP_003735339.1 | 1 - 2 | 984 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGLN1| Gene Symbol: EGLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735291.3 | XP_003735339.1 | 1 - 2 | 984 | 66 | |
Set Membership: |
|