Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748193.4 | XP_002748239.3 | 1 - 2 | 312 | 87 | |
| XM_008996996.2 | XP_008995244.2 | 1 - 2 | 316 | 87 | ||
| XM_008996998.2 | XP_008995246.2 | 1 - 2 | 318 | 87 | ||
More Information
Set Membership: |
|

DHRS11
Genomic Map