Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748293.3 | XP_002748339.1 | 2 - 3 | 1043 | 100 | |
| XM_008997260.2 | XP_008995508.1 | 2 - 3 | 482 | 100 | ||
More Information
Set Membership: |
|

SPAG7
Genomic Map