Assay Details
| Gene Symbol: SLC25A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009827.2 | XP_009008075.1 | 3 - 4 | 516 | 72 | |
| XM_017979063.1 | XP_017834552.1 | 3 - 4 | 516 | 72 | ||
| XM_017979064.1 | XP_017834553.1 | 3 - 4 | 516 | 72 | ||
More Information
Set Membership: |
|

SLC25A16
Genomic Map