Assay Details
Gene Symbol: NHSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017968536.1 | XP_017824025.1 | 3 - 4 | 3754 | 70 | |
XM_017968537.1 | XP_017824026.1 | 3 - 4 | 3993 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
