Assay Details
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979245.2 | XP_008977493.1 | 14 - 15 | 4476 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLC1| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979245.2 | XP_008977493.1 | 14 - 15 | 4476 | 95 | |
Set Membership: |
|