Assay Details
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205139.2 | NP_001192068.2 | 6 - 7 | 618 | 60 | |
NM_001278383.1 | NP_001265312.1 | 6 - 7 | 621 | 60 | ||
GenBank mRNA | HX584077.1 | - | 5 - 6 | 371 | 60 | |
HX652217.1 | - | 6 - 7 | 619 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
