Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998586.1 | XP_008996834.1 | 4 - 5 | 1274 | 95 | |
| XM_008998633.1 | XP_008996881.1 | 4 - 5 | 1161 | 95 | ||
| XM_008998680.2 | XP_008996928.1 | 4 - 5 | 827 | 95 | ||
| XM_008998730.2 | XP_008996978.1 | 4 - 5 | 823 | 95 | ||
| XM_008998769.2 | XP_008997017.1 | 4 - 5 | 826 | 95 | ||
| XM_008998870.1 | XP_008997118.1 | 4 - 5 | 1274 | 95 | ||
| XM_017970358.1 | XP_017825847.1 | 3 - 4 | 1304 | 95 | ||
More Information
Set Membership: |
|

BIVM
Genomic Map