Assay Details
| Gene Symbol: INSYN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756710.3 | XP_002756756.1 | 2 - 3 | 1343 | 92 | |
| XM_017963272.1 | XP_017818761.1 | 1 - 2 | 2771 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSYN2| Gene Symbol: INSYN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756710.3 | XP_002756756.1 | 2 - 3 | 1343 | 92 | |
| XM_017963272.1 | XP_017818761.1 | 1 - 2 | 2771 | 92 | ||