Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017965651.1 | XP_017821140.1 | 3 - 4 | 514 | 80 | |
XM_017965652.1 | XP_017821141.1 | 2 - 3 | 406 | 80 | ||
XM_017965653.1 | XP_017821142.1 | 3 - 4 | 514 | 80 | ||
XM_017965654.1 | XP_017821143.1 | 2 - 3 | 406 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
