Assay Details
| Gene Symbol: SS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757130.3 | XP_002757176.2 | 4 - 5 | 713 | 63 | |
| XM_002757131.4 | XP_002757177.1 | 4 - 5 | 713 | 63 | ||
| XM_017963934.1 | XP_017819423.1 | 4 - 5 | 713 | 63 | ||
| XM_017963935.1 | XP_017819424.1 | 4 - 5 | 435 | 63 | ||
More Information
Set Membership: |
|

SS18
Genomic Map