Assay Details
Gene Symbol: LOC100390879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749792.2 | XP_002749838.1 | 5 - 6 | 855 | 88 | |
XM_008999492.2 | XP_008997740.1 | 4 - 5 | 1710 | 88 | ||
XM_008999494.2 | XP_008997742.1 | 6 - 7 | 895 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
