Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 7 - 8 | 868 | 111 | |
XM_008988341.2 | XP_008986589.1 | 7 - 8 | 1026 | 111 | ||
XM_008988348.2 | XP_008986596.1 | 8 - 9 | 943 | 111 | ||
XM_008988357.2 | XP_008986605.1 | 6 - 7 | 745 | 111 | ||
XM_017969030.1 | XP_017824519.1 | 6 - 7 | 1325 | 111 | ||
XM_017969035.1 | XP_017824524.1 | 6 - 7 | 1326 | 111 | ||
XM_017969036.1 | XP_017824525.1 | 4 - 5 | 1146 | 111 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
