Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 9 - 10 | 1138 | 92 | |
XM_008988341.2 | XP_008986589.1 | 9 - 10 | 1296 | 92 | ||
XM_008988348.2 | XP_008986596.1 | 10 - 11 | 1213 | 92 | ||
XM_008988357.2 | XP_008986605.1 | 8 - 9 | 1015 | 92 | ||
XM_017969030.1 | XP_017824519.1 | 8 - 9 | 1595 | 92 | ||
XM_017969035.1 | XP_017824524.1 | 8 - 9 | 1596 | 92 | ||
XM_017969036.1 | XP_017824525.1 | 6 - 7 | 1416 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
