Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745987.4 | XP_002746033.3 | 3 - 4 | 814 | 67 | |
| XM_017970662.1 | XP_017826151.1 | 3 - 4 | 841 | 67 | ||
More Information
Set Membership: |
|

BST1
Genomic Map