Assay Details
| Gene Symbol: ICAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967944.1 | XP_017823433.1 | 2 - 3 | 714 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ICAM5| Gene Symbol: ICAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967944.1 | XP_017823433.1 | 2 - 3 | 714 | 100 | |
Set Membership: |
|