Assay Details
Gene Symbol: COASY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003732907.3 | XP_003732955.2 | 8 - 9 | 1749 | 67 | |
XM_008996643.1 | XP_008994891.1 | 8 - 9 | 1656 | 67 | ||
XM_017972360.1 | XP_017827849.1 | 8 - 9 | 1653 | 67 | ||
GenBank mRNA | EF214992.1 | - | 3 - 4 | 198 | 67 | |
EF215159.1 | - | 3 - 4 | 208 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
