Assay Details
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017967581.1 | XP_017823070.1 | 6 - 7 | 846 | 87 | |
XM_017967582.1 | XP_017823071.1 | 5 - 6 | 1431 | 87 | ||
XM_017967583.1 | XP_017823072.1 | 6 - 7 | 831 | 87 | ||
XM_017967584.1 | XP_017823073.1 | 5 - 6 | 1428 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
