Assay Details
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754496.3 | XP_002754542.1 | 2 - 3 | 397 | 64 | |
XM_002754497.4 | XP_002754543.2 | 2 - 3 | 397 | 64 | ||
XM_002754498.2 | XP_002754544.1 | 2 - 3 | 397 | 64 | ||
XM_009006846.2 | XP_009005094.1 | 2 - 3 | 397 | 64 | ||
XM_009006847.2 | XP_009005095.1 | 2 - 3 | 397 | 64 | ||
XM_009006848.2 | XP_009005096.1 | 2 - 3 | 397 | 64 | ||
XM_017977552.1 | XP_017833041.1 | 2 - 3 | 2860 | 64 | ||
XM_017977553.1 | XP_017833042.1 | 2 - 3 | 397 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
