Assay Details
Gene Symbol: SEPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748771.3 | XP_002748817.1 | 6 - 7 | 1815 | 84 | |
XM_008997807.2 | XP_008996055.1 | 6 - 7 | 2000 | 84 | ||
XM_008997808.2 | XP_008996056.1 | 6 - 7 | 1304 | 84 | ||
XM_008997809.2 | XP_008996057.1 | 6 - 7 | 1897 | 84 | ||
XM_008997810.2 | XP_008996058.1 | 5 - 6 | 1103 | 84 | ||
XM_008997811.2 | XP_008996059.1 | 5 - 6 | 696 | 84 | ||
XM_008997813.2 | XP_008996061.1 | 5 - 6 | 554 | 84 | ||
XM_008997816.2 | XP_008996064.1 | 5 - 6 | 556 | 84 | ||
XM_017972879.1 | XP_017828368.1 | 5 - 6 | 903 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
