Assay Details
| Gene Symbol: NAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995849.2 | XP_008994097.1 | 6 - 7 | 593 | 102 | |
| XM_008995850.1 | XP_008994098.1 | 3 - 4 | 199 | 102 | ||
| XM_017972013.1 | XP_017827502.1 | 6 - 7 | 861 | 102 | ||
More Information
Set Membership: |
|

NAPB
Genomic Map