Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747063.3 | XP_002747109.1 | 2 - 3 | 2146 | 98 | |
XM_017971457.1 | XP_017826946.1 | 2 - 3 | 2146 | 98 | ||
XM_017971458.1 | XP_017826947.1 | 2 - 3 | 2146 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
