Assay Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758720.3 | XP_002758766.1 | 5 - 6 | 945 | 73 | |
| XM_008982297.2 | XP_008980545.1 | 5 - 6 | 776 | 73 | ||
| XM_008982298.2 | XP_008980546.1 | 5 - 6 | 1365 | 73 | ||
| XM_017965163.1 | XP_017820652.1 | 5 - 6 | 3082 | 73 | ||
| XM_017965164.1 | XP_017820653.1 | 5 - 6 | 1312 | 73 | ||
| XM_017965165.1 | XP_017820654.1 | 5 - 6 | 711 | 73 | ||
More Information
Set Membership: |
|

ZNF148
Genomic Map