Assay Details
| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759628.4 | XP_002759674.1 | 2 - 3 | 465 | 82 | |
| XM_008983852.2 | XP_008982100.1 | 2 - 3 | 465 | 82 | ||
| XM_008983853.2 | XP_008982101.1 | 2 - 3 | 465 | 82 | ||
| XM_017965873.1 | XP_017821362.1 | 2 - 3 | 465 | 82 | ||
More Information
Set Membership: |
|

METTL6
Genomic Map