Assay Details
Gene Symbol: LOC103787289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009008361.2 | XP_009006609.1 | 3 - 4 | 280 | 105 | |
XM_017978217.1 | XP_017833706.1 | 3 - 4 | 283 | 105 | ||
XM_017978218.1 | XP_017833707.1 | 3 - 4 | 191 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
