Assay Details
| Gene Symbol: RBPMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756918.3 | XP_002756964.2 | 4 - 5 | 833 | 71 | |
| XM_008979259.2 | XP_008977507.1 | 4 - 5 | 211 | 71 | ||
| XM_008979260.2 | XP_008977508.1 | 4 - 5 | 833 | 71 | ||
| XM_008979264.2 | XP_008977512.1 | 4 - 5 | 219 | 71 | ||
| XM_017963752.1 | XP_017819241.1 | 4 - 5 | 833 | 71 | ||
| XM_017963753.1 | XP_017819242.1 | 4 - 5 | 833 | 71 | ||
| XM_017963754.1 | XP_017819243.1 | 4 - 5 | 833 | 71 | ||
| XM_017963755.1 | XP_017819244.1 | 4 - 5 | 833 | 71 | ||
| XM_017963757.1 | XP_017819246.1 | 4 - 5 | 833 | 71 | ||
More Information
Set Membership: |
|

RBPMS
Genomic Map