Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270906.1 | NP_001257835.1 | 1 - 2 | 346 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270906.1 transcripts.
Set Membership: |
|

ZNF460
Genomic Map