Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270906.1 | NP_001257835.1 | 2 - 3 | 469 | 61 | |
| XM_008988742.2 | XP_008986990.1 | 2 - 3 | 678 | 61 | ||
More Information
Set Membership: |
|

ZNF460
Genomic Map