Assay Details
| Gene Symbol: NUDT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743028.2 | XP_002743074.1 | 2 - 3 | 282 | 80 | |
| XM_003731998.3 | XP_003732046.1 | 2 - 3 | 700 | 80 | ||
| XM_003731999.2 | XP_003732047.1 | 2 - 3 | 411 | 80 | ||
More Information
Set Membership: |
|

NUDT2
Genomic Map