Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759806.4 | XP_002759852.1 | 2 - 3 | 449 | 73 | |
XM_017966184.1 | XP_017821673.1 | 2 - 3 | 385 | 73 | ||
XM_017966185.1 | XP_017821674.1 | 1 - 2 | 316 | 73 | ||
XM_017966186.1 | XP_017821675.1 | 2 - 3 | 430 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017966186.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
