Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759806.4 | XP_002759852.1 | 3 - 4 | 999 | 76 | |
XM_008984173.2 | XP_008982421.1 | 2 - 3 | 885 | 76 | ||
XM_017966184.1 | XP_017821673.1 | 3 - 4 | 935 | 76 | ||
XM_017966185.1 | XP_017821674.1 | 2 - 3 | 866 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
