Assay Details
Gene Symbol: PMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760094.3 | XP_002760140.1 | 4 - 5 | 735 | 56 | |
XM_008984595.2 | XP_008982843.1 | 4 - 5 | 2429 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
