Assay Details
| Gene Symbol: ACOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735703.3 | XP_003735751.1 | 13 - 14 | 1173 | 62 | |
| XM_008989078.2 | XP_008987326.1 | 12 - 13 | 1176 | 62 | ||
More Information
Set Membership: |
|

ACOT9
Genomic Map