Assay Details
| Gene Symbol: LOC100390841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732618.3 | XP_003732666.3 | 4 - 5 | 2835 | 78 | |
| XM_017970953.1 | XP_017826442.1 | 4 - 5 | 2840 | 78 | ||
| XM_017970954.1 | XP_017826443.1 | 4 - 5 | 2810 | 78 | ||
More Information
Set Membership: |
|

LOC100390841
Genomic Map