Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204801.2 | NP_001191730.2 | 3 - 4 | 409 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FPGT| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204801.2 | NP_001191730.2 | 3 - 4 | 409 | 107 | |
Set Membership: |
|