Assay Details
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008999945.2 | XP_008998193.2 | 9 | 3288 | 99 | |
XM_008999946.2 | XP_008998194.1 | 9 | 3285 | 99 | ||
XM_008999951.2 | XP_008998199.1 | 9 | 4249 | 99 | ||
XM_008999952.2 | XP_008998200.2 | 8 | 3204 | 99 | ||
XM_008999953.2 | XP_008998201.1 | 9 | 3413 | 99 | ||
XM_017974065.1 | XP_017829554.1 | 8 | 3167 | 99 | ||
XM_017974066.1 | XP_017829555.1 | 8 | 4166 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
