Assay Details
| Gene Symbol: PHETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004630.2 | XP_009002878.1 | 3 | 1820 | 73 | |
| XM_017976379.1 | XP_017831868.1 | 3 | 2165 | 73 | ||
More Information
Set Membership: |
|

PHETA1
Genomic Map