Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745231.3 | XP_002745277.2 | 2 - 3 | 1002 | 91 | |
XM_008992311.2 | XP_008990559.1 | 2 - 3 | 1001 | 91 | ||
XM_008992312.2 | XP_008990560.1 | 3 - 4 | 1125 | 91 | ||
GenBank mRNA | EU488858.1 | - | 1 - 2 | 387 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
