Assay Details
Gene Symbol: RNLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756369.3 | XP_002756415.1 | 4 - 5 | 1906 | 68 | |
XM_017979119.1 | XP_017834608.1 | 4 - 5 | 1931 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
