Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750852.3 | XP_002750898.1 | 7 - 8 | 1245 | 65 | |
XM_009001277.1 | XP_008999525.1 | 8 - 9 | 1348 | 65 | ||
XM_009001278.2 | XP_008999526.1 | 8 - 9 | 1426 | 65 | ||
XM_009001279.1 | XP_008999527.1 | 7 - 8 | 1216 | 65 | ||
XM_009001280.1 | XP_008999528.1 | 6 - 7 | 1099 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
