Assay Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761138.4 | XP_002761184.1 | 8 - 9 | 939 | 76 | |
XM_008986195.2 | XP_008984443.1 | 8 - 9 | 825 | 76 | ||
XM_008986196.2 | XP_008984444.1 | 8 - 9 | 888 | 76 | ||
XM_008986197.2 | XP_008984445.1 | 8 - 9 | 1029 | 76 | ||
XM_017966827.1 | XP_017822316.1 | 8 - 9 | 929 | 76 | ||
XM_017966828.1 | XP_017822317.1 | 8 - 9 | 777 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
