Assay Details
| Gene Symbol: KEAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735487.3 | XP_003735535.1 | 3 - 4 | 1495 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KEAP1| Gene Symbol: KEAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735487.3 | XP_003735535.1 | 3 - 4 | 1495 | 61 | |
Set Membership: |
|