Assay Details
| Gene Symbol: CYB5R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743813.4 | XP_002743859.1 | 2 - 3 | 389 | 69 | |
| XM_017963941.1 | XP_017819430.1 | 2 - 3 | 282 | 69 | ||
| XM_017963942.1 | XP_017819431.1 | 2 - 3 | 267 | 69 | ||
More Information
Set Membership: |
|

CYB5R3
Genomic Map