Assay Details
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755885.4 | XP_002755931.1 | 3 - 4 | 369 | 67 | |
XM_009009154.2 | XP_009007402.1 | 3 - 4 | 398 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
