Assay Details
Gene Symbol: LOC100390879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749792.2 | XP_002749838.1 | 2 - 3 | 455 | 61 | |
XM_008999492.2 | XP_008997740.1 | 1 - 2 | 1310 | 61 | ||
XM_008999494.2 | XP_008997742.1 | 3 - 4 | 495 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
