Assay Details
| Gene Symbol: CRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761630.4 | XP_002761676.1 | 3 - 4 | 712 | 88 | |
| XM_008987068.2 | XP_008985316.1 | 3 - 4 | 695 | 88 | ||
More Information
Set Membership: |
|

CRB3
Genomic Map