Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 7 - 8 | 2543 | 87 | |
XM_009009599.2 | XP_009007847.1 | 8 - 9 | 2536 | 87 | ||
XM_009009601.2 | XP_009007849.1 | 8 - 9 | 2536 | 87 | ||
XM_009009602.2 | XP_009007850.1 | 7 - 8 | 2329 | 87 | ||
XM_009009603.2 | XP_009007851.1 | 7 - 8 | 1012 | 87 | ||
XM_017978940.1 | XP_017834429.1 | 9 - 10 | 2612 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
