Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 8 - 9 | 2662 | 60 | |
XM_009009599.2 | XP_009007847.1 | 9 - 10 | 2655 | 60 | ||
XM_009009602.2 | XP_009007850.1 | 8 - 9 | 2448 | 60 | ||
XM_009009603.2 | XP_009007851.1 | 8 - 9 | 1131 | 60 | ||
XM_017978940.1 | XP_017834429.1 | 10 - 11 | 2731 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
