Assay Details
| Gene Symbol: PCMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982617.2 | XP_008980865.1 | 6 - 7 | 865 | 86 | |
| XM_008982619.2 | XP_008980867.1 | 4 - 5 | 597 | 86 | ||
| XM_017965335.1 | XP_017820824.1 | 7 - 8 | 1111 | 86 | ||
| XM_017965336.1 | XP_017820825.1 | 4 - 5 | 588 | 86 | ||
More Information
Set Membership: |
|

PCMTD1
Genomic Map