Assay Details
Gene Symbol: FAM198B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745288.3 | XP_002745334.1 | 2 - 3 | 1515 | 82 | |
XM_008992543.2 | XP_008990791.1 | 2 - 3 | 1313 | 82 | ||
XM_008992544.2 | XP_008990792.1 | 2 - 3 | 1406 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
